missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ Custom 5' Fluorescent Labeled/Unlabeled Pairs
Description
A 5' fluorescent dye-labeled oligo and unlabeled complement are supplied in separate tubes. Choose among 6-FAM™, TET™, VIC™, HEX™, NED™, and PET™ as labels. Ideal for use in fragment analysis, gene expression studies, genotyping, and PCR. Oligos are desalted.
Specifications
Specifications
| Product Type | Oligonucleotides |
| Length (Metric) | 1.524 m |
| Content And Storage | Store in Freezer (-5°C to -30°C). |
| Purification Method | Desalted |
| Label or Dye | 6-FAM, HEX, NED, PET, TET, VIC |
| Primer | 5' Fluorescent Labeled/Unlabeled |
| Quantity | 300,000 pmol⁄each |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?