missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Custom 5' Fluorescent Labeled/Unlabeled Pairs

Product Code. 15597240 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
10,000 pmol
300,000 pmol
80,000 pmol
Unit Size:
10000pmol
300000pmol
80000pmol
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15597240 80,000 pmol 80000pmol
15557240 10,000 pmol 10000pmol
15577280 300,000 pmol 300000pmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15597240 Supplier Applied Biosystems™ Supplier No. 4304981
 Restricted Item
Estimated Shipment: 18-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Custom 5' Fluorescent Labeled/Unlabeled Di-repeat + Tail

A 5' fluorescent dye-labeled oligo and unlabeled complement are supplied in separate tubes. Choose among 6-FAM™, TET™, VIC™, HEX™, NED™, and PET™ as labels. Ideal for use in fragment analysis, gene expression studies, genotyping, and PCR. Oligos are desalted.
TRUSTED_SUSTAINABILITY

Specifications

Description Custom 5' Labeled/Unlabeled Di-Repeat + Tail, 80000 pmol, 6-FAM™, TET™, VIC™, HEX™, NED™, PET™ 5' Reporter Dye, Choice of Delivered Scales, These Products can be used in a Variety of Applications
Product Type Custom 5' Labeled or Unlabeled Di-Repeat + Tail
Content And Storage -20°C
Quantity 80,000 pmol
For Use With (Application) Fragment Analysis, Gene Expression Studies, Genotyping, PCR
Form Delivered either in Solution or Dried down

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.