missing translation for 'onlineSavingsMsg'
Learn More

Value DNA Oligo , Desalted, Liquid, < 24 , 25 nmol

Product Code. 15460704
Change view
Click to view available options
Quantity:
25 nmol
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15460704 25 nmol Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15460704 Supplier Invitrogen Supplier No. A15611

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Custom DNA Oligo, 25N, Desalted, Liquid

Value Oligos are the most cost-effective and fastest way to order oligonucleotides from us. Simply enter your sequence and then check out. Value Oligos are available at 25 nmol or 50 nmol scales, with a range of purification options to suit your needs.
TRUSTED_SUSTAINABILITY

Specifications

Description DNA Oligo, 25 nmole, 25N, Liquid, Desalted, Cost-effective and Fastest
Product Type DNA Oligo
Purification Method Desalted
Quantity 25 nmol
Form Liquid
Product Title
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.