missing translation for 'onlineSavingsMsg'
Learn More

Value DNA Oligo , HPLC, Liquid, < 24 , 50 nmol

Product Code. 15430704
Change view
Click to view available options
Quantity:
50 nmol
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15430704 50 nmol Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15430704 Supplier Invitrogen Supplier No. A15606
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Custom DNA Oligo, 50N, HPLC, Liquid

Value Oligos are the most cost-effective and fastest way to order oligonucleotides from us. Simply enter your sequence and then check out. Value Oligos are available at 25 nmol or 50 nmol scales, with a range of purification options to suit your needs.

TRUSTED_SUSTAINABILITY

Specifications

Description DNA Oligo, 50 nmol, 50N, HPLC Purity, Liquid
Product Type Custom DNA Oligo
Quantity 50 nmol
Grade HPLC
Form Liquid
Can I order a primer with degenerate bases?

Yes, we can synthesize primers with degenerate bases. Please see here for the electronic ordering code to use (https://www.thermofisher.com/us/en/home/products-and-services/product-types/primers-oligos-nucleotides/invitrogen-custom-dna-oligos/oligo-ordering-details/oligo-modification-options.html#2).

Do you offer TE buffer that I can use to resuspend my oligos?

We offer TE Buffer, pH 8.0 (Cat. Nos. AM9849 and AM9858), that consists of 10 mM Tris (adjusted to pH 8.0 with HCl) and 1 mM EDTA.

I am designing my own primers. Will they be shipped to me as lyophilized primers or in solution?

Unless you specifically request that primers be supplied in solution, they are shipped lyophilized. We recommend reconstituting them in the appropriate volume of TE (10 mM Tris-HCl, 1 mM EDTA, pH 8.0).

If I do not specify a modification for my primer, will the primer have a 5‘ phosphate?

No, the primer will not have this modification. You must specify that it should have a 5‘ phosphate modification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.