missing translation for 'onlineSavingsMsg'
Learn More

Custom PreAmp Pools

Product Code. 15504344 Shop All Applied Biosystems Products
Change view
Click to view available options
Format:
96A
96B
Format 12
Format 16
Format 24
Format 32
Format 384
Format 48
Format 64
Unit Size:
1mL
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Format unitSize
15504344 Format 12 1mL
15514344 Format 16 1mL
15524344 Format 24 1mL
15534344 Format 32 1mL
15544344 Format 48 1mL
15554344 Format 64 1mL
15564344 96A 1mL
15574344 96B 1mL
15504364 Format 384 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 15504344 Supplier ABI Applied Biosystems Supplier No. 4459655
  • ONLINE EXCLUSIVE PRICE
    478.00 €
    391.96 € / 1mL
    Save 86.04 €
    18% Off
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Custom PreAmp Pools

Custom miRNA array designs can hold from 12 to 380 assays, and 1 to 8 samples on each array design. Ten formats are offered, available only with microfluidics card design. Use Custom PreAmp pools for samples with limited miRNA amounts.
TRUSTED_SUSTAINABILITY

Specifications

Type Custom TaqMan™ miRNA Pre-Amp Pool
Format Format 12
Content And Storage Room Temperature
No. of Samples 1 to 8
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.