Learn More
Custom Primer
Product Code. 10184553
Product Code. 10184553 Supplier Invitrogen Supplier No. 11739026
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10 μmole.
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10 μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).