Learn More
Custom Primer
Product Code. 10357843
Product Code. 10357843 Supplier Invitrogen Supplier No. 11772027
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10 μmole.
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10 μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).