Learn More
Custom RNA
Product Code. 10572504
Change view
Click to view available options
Product Type:
Custom RNA, HPLC, 1.7μmol
Custom RNA, HPLC, 1 μmol
Custom RNA, HPLC, 200nmol
Unit Size:
1.7µmol
1µmol
200nmol
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
10572504 Custom RNA, HPLC, 1 μmol 1µmol
10174082 Custom RNA, HPLC, 200nmol 200nmol
10388863 Custom RNA, HPLC, 1.7μmol 1.7µmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10572504 Supplier Invitrogen Supplier No. 10620105
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Using our award-winning BLOCK-iT™ RNAi Designer, you can design Custom BLOCK-iT™ siRNA sequences for efficient and specific gene expression knockdowns in any organism
  • Using our award-winning BLOCK-iT™ RNAi Designer, you can design Custom BLOCK-iT™ siRNA sequences for efficient and specific gene expression knockdowns in any organism.
  • For enhanced target specificity and stability in serum, Stealth RNAi™ RNAi chemistry is available.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Custom RNA, HPLC, 1 μmol
Product Title
Select an issue
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Datenschutzrichtlinie.