missing translation for 'onlineSavingsMsg'
Learn More

Ambion™ Custom RNA, Stealth, HPLC

Product Code. 10667923
Change view
Click to view available options
Product Type:
RNA HPLC In-vivo
Stealth RNA HPLC
Quantity:
20 nmol
5 nmol
500 nmol
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Quantity Unit Size
10667923 RNA HPLC In-vivo 500 nmol Each
10164353 Stealth RNA HPLC 20 nmol Each
10523054 RNA HPLC In-vivo 20 nmol Each
10328033 Stealth RNA HPLC 500 nmol Each
10522504 Stealth RNA HPLC 5 nmol Each
10174283 RNA HPLC In-vivo 5 nmol Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 10667923 Supplier Ambion™ Supplier No. 100001865
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Custom RNA, Stealth, HPLC

Use this tool for fast and easy online ordering of bench-tested Validated Stealth RNAi™ siRNA, ready-to-order Stealth Select RNAi™ siRNA, bench-tested BLOCK-iT™ Pol II miR Validated miRNA Vector Duopaks, and ready-to-clone BLOCK-iT™ miR RNAi Select.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Room temperature
Description RNA HPLC In-vivo, 500nmol, Lowest Off-target Effects and Highest Potency, Small, Chemically Modified Single-Stranded RNA Molecules Designed to Specifically Inhibit Endogenous miRNAs for the Purpose of Functional Analysis Via Down-Regulation
Product Type RNA HPLC In-vivo
Purity HPLC
Quantity 500 nmol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.