missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Custom TaqMan™ Gene Expression Assay, VIC
GREENER_CHOICE

Product Code. 15819854 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
L (2900 reactions), made to order
M (750 reactions), made to order
S (360 reactions), made to order
Unit Size:
2900 reactions
360 reactions
750 reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15819854 S (360 reactions), made to order 360 reactions
15839854 L (2900 reactions), made to order 2900 reactions
15829854 M (750 reactions), made to order 750 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15819854 Supplier Applied Biosystems™ Supplier No. 4448508
Simplify Your Gene Expression Workflow
Use promo code 27975
View all promotions
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Applied Biosystems Custom TaqMan Gene Expression assays, VIC, are the custom version of our predesigned TaqMan assays for quantitative real-time PCR.

Applied Biosystems Custom TaqMan Gene Expression assays, VIC, are the custom version of our predesigned TaqMan assays for quantitative real-time PCR. Custom TaqMan Gene Expression assays consist of a pair of unlabeled PCR primers and a TaqMan probe with a dye label (VIC) on the 5’ end and a minor groove binder (MGB) and nonfluorescent quencher (NFQ) on the 3’ end.

Features include:
Easy to use—just add cDNA and master mix and run the qPCR—no melt curves required
Specific—TaqMan assays use proprietary MGB-containing probes that can be up to 15 bases shorter than non-MGB probes, improving the specificity of the assay
Sensitive—TaqMan assays are ideal for measuring low levels of expression or low-abundance targets
Accurate—identify small fold-changes with high accuracy of quantitation
Gold-standard TaqMan qPCR chemistry—TaqMan assays draw on Thermo Fisher Scientific's bioinformatics assay design pipeline to help ensure high specificity and minimal cross-reactivity, even for gene variants with high sequence homology
Ideal for multiplexing—combine one FAM-labeled and one VIC-labeled assay to create a duplex assay for two different gene targets in the same qPCR well
Sequences provided—with every Custom TaqMan Gene Expression you design and order

Approximate ship time
Made to order: 4–6 days in North America, 6–10 days in Europe

Pre-formulated assay
• 2 unlabeled PCR primers (900 nM each final 1X concentration)
• 1 VIC dye-labeled TaqMan MGB probe (250 nM final 1X concentration)

TaqMan Gene Expression assays are the gold standard in real-time PCR gene expression studies, built on more than 20 years of experience. Each assay includes target primers and a sequence-specific probe optimized for the best functional performance. No additional design, optimization, or melt curve analysis is needed. Available in a wide variety of formats and species, new assay designs are constantly added to help meet your research needs. TaqMan assays have been cited in over 40,000 publications and are backed by more than 350 patents.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 20X
Content And Storage 1 Tube containing a 20 x (S and M sizes) or 60 x (L size) mix of pre-formulated assay (1 probe and 2 primers).

Store (at -15°C to -25°C.

)
For Use With (Equipment) 7500 System, 7500 Fast System, 7900HT System, StepOne™, StepOnePlus™, ViiA™ 7 System, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio™ 12k Flex
Primer Labeled PCR
Product Line TaqMan
Product Type Gene Expression Assay
Quantity S (360 reactions), made to order
Shipping Condition Room Temperature
No. of Reactions 360 reactions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.