missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ TAMRA Probe
GREENER_CHOICE

Product Code. 11529436
Click to view available options
Quantity:
20,000 pmol
50,000 pmol
6,000 pmol
Product Type:
TAMRA Probe
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11529436

Brand: Applied Biosystems™ 450025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Custom TaqMan™ Probe

Custom TaqMan™ probes are dual-labeled primers suitable for use in any real-time PCR applicaton. They incorporate a 5' reporter dye and a 3' TAMRA™ nonfluorescent quencher. Choose among FAM™, VIC™, ABY™, and JUN™ as 3' reporters. Probes are HPLC-purified.
TRUSTED_SUSTAINABILITY

Specifications

3'Primer Modification TAMRA
5'Primer Modification FAM, TET, VIC
Content And Storage 1 tube containing a dual-labeled probe in solution or dried down.

Store at -15°C to -25°C
Form Liquid or Dried Down
Format Tube
For Use With (Equipment) 7500 System, 7500 Fast System, 7900HT System, StepOne™, StepOnePlus™, ViiA™ 7 System, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio™ 12k Flex
Green Features Sustainable packaging
Internal Probe Modification None
Product Line TaqMan
Product Type TAMRA Probe
Purification Method HPLC
Purity or Quality Grade HPLC
Quantity 6,000 pmol
Shipping Condition Room Temperature
For Use With (Application) Real Time PCR (qPCR)
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.