missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Custom TaqMan™ Small RNA Assay

Product Code. 15514334 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
L (2900 RT/2900 PCR reactions), made to order
M (750 RT/750 PCR reactions), made to order
S (50 RT/150 PCR reactions), made to order
XS (25 RT/75 PCR reactions), made to order
Unit Size:
2900 reactions
75 reactions
750 reactions
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15514334 XS (25 RT/75 PCR reactions), made to order 75 reactions
15449142 S (50 RT/150 PCR reactions), made to order Each
15581044 M (750 RT/750 PCR reactions), made to order 750 reactions
15591044 L (2900 RT/2900 PCR reactions), made to order 2900 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Product Code. 15514334 Supplier Applied Biosystems™ Supplier No. 4440418

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price

Custom TaqMan Small RNA Assays use TaqMan Real-Time PCR Assays for the analysis of any small RNA molecule.

Custom TaqMan Small RNA Assays use TaqMan Real-Time PCR Assays for the analysis of any small RNA molecule. The novel adaptations in TaqMan Assay design developed for the study of miRNAs are ideal for analysis of any small nucleic acid less than 200 bases long, including newly discovered miRNAs, Piwi-interacting RNA (piRNA), small nuclear RNA (snRNA), and small nucleolar RNA (snoRNA).

Flexible and convenient ordering—easy process with our secure Custom TaqMan Small RNA Assay design tool
Versatile application—use for the analysis of any small RNA from any species
Reliable outcome—pre-formulated assays are optimized to run under universal conditions
Powerful miRNA quantitation—the same sensitivity, specificity, and reproducibility as predesigned TaqMan MicroRNA Assays

Sophisticated design pipeline
The miRPipe Small RNA Assay design pipeline is founded on our extensive background and bioinformatics expertise designing TaqMan Gene Expression Assays. To detect very small targets, it includes design features based on empirical data gathered over years of experiments. The resulting automated pipeline includes built-in flexibility features enabling design of assays for the broadest assortment of small RNA sequences. The benefits of TaqMan Assay technology are ideal for analysis of virtually any small RNA, enabling results you can trust from the very first experiment.

Pre-formulated assay
• Forward primer: 1500 nM
• Reverse primer: 700 nM
• FAM-MGB probe: 200 nM
• RT sequences: 50 nM

Ordering information
First, bioinformatically qualify your sequence target for specificity and sequence quality, then use the Custom TaqMan Small RNA Assay Design Tool to input your sequence and submit it for assay design. This easy-to-use tool is a secure and confidential. If we are unable to design or manufacture an assay for your sequence target of interest, you will not be charged.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 20X (Assay), 5X (RT Primer)
Content And Storage • 1 tube containing RT primer at concentration of 5X (XS and S sizes), 20X (M size), or 60X (L size)
• 1 tube containing a 20X (XS, S, and M sizes) or 60X (L size) mix of pre-formulated assay (1 probe and 2 primers).

Store at -15 to -25°C.

Form Liquid
Product Line TaqMan
Quantity XS (25 RT/75 PCR reactions), made to order
Shipping Condition Room Temperature
For Use With (Application) Gene Expression
No. of Reactions 25 reactions (PCR), 75 reactions (RT-PCR)

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.