Learn More
SHOWA Cut and Abrasion-Resistant Knit Gloves
Product Code. 17805532
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Unit Size:
10 pairs
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17805532 Medium 10 pairs
17825532 Small 10 pairs
17895522 Large 10 pairs
17835532 X-Large 10 pairs
17815532 2X-Large 10 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 17805532 Supplier SHOWA Supplier No. 54207M
In Stock
Add to basket
Edge
Add to basket
The 542 cut and abrasion-resistant glove safeguards your hands while working with components or in hazardous situations. Its polyurethane coating over a reinforced HPPE liner shields against abrasion and oils while allowing for maximum flexibility and dexterity.
  • The SHOWA 542 cut and abrasion-resistant glove is a high-quality protective glove that is designed to keep your hands safe during hazardous applications and component assembly tasks.
  • This glove features a unique combination of materials that provide superior protection against cuts, abrasion, and oils, while still maintaining excellent flexibility and dexterity.
  • The polyurethane coating enhances the glove's grip, allowing you to work with small components or slippery materials with ease. Additionally, the HPPE reinforced liner provides excellent cut resistance, making this glove ideal for handling sharp objects.
  • Designed with comfort in mind. The open back of hand design allows air to circulate around your hands, reducing perspiration and keeping your hands cool and comfortable. The seamless liner minimizes irritation and friction, even during extended wear periods, so you can keep your hands protected without sacrificing comfort.
  • The unique combination of materials allows for maximum flexibility, so you can easily handle small components or perform intricate tasks without feeling restricted.
  • The SHOWA 542 glove is available in a range of sizes, making it easy to find the perfect fit for your hands. Whether you're working in an industrial setting, handling sharp objects, or working with hazardous materials, the SHOWA 542 glove provides the protection and comfort you need to get the job done safely and efficiently.
Specifications
Construction Knit
Coating Material Polyurethane
Color White
Hand Specific
Latex Free Yes
Material Polyurethane
Product Type Cut- and Abrasion-resistant Glove
Size Medium
EN388 Standard EN388:4341B
Gender Unisex
Glove Cuff Style Elasticated
Model 542
Puncture Rating ASTM F2878 LEVEL 3