missing translation for 'onlineSavingsMsg'
Learn More

Hach Lange™ Cuvette Tests

Product Code. 10594493
Change view
Click to view available options
Type:
Ammonium Cuvette Test
Phosphate Ortho/Total Cuvette Test
Unit Size:
Each
Pack of 25
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
10594493 Ammonium Cuvette Test Each
10619741 Phosphate Ortho/Total Cuvette Test Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10594493 Supplier Hach Lange™ Supplier No. LCK304
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

For use with water analysis, these exemplary cuvette tests have ISO 6878-1-1986 and DIN 38405 D11-4 certifications. They utilize the Phosphormolybdenum blue testing method for the DR3900, DR6000, DR1900, DR2800, DR3800, DR5000 instruments.

  • Ammonium cuvette test 0.015-2.0 mg/L NH4-N
  • ISO 7150-1, DIN 38406 E5-1, UNI 11669:2017 certifications
  • DR3900, DR6000, DR1900, DR2800, DR3800, DR5000 instruments
  • Indophenol Blue methods
  • Precise and reliable measurement values
  • 0.015 – 2.0 mg/L NH4-N measuring range 
  • 0.02 - 2.5 mg/L NH4 measuring range (2)
  • Has a shelf life of 17 months from production date
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO 7150-1, DIN 38406 E5-1
Color White
For Use With (Application) Water analysis
For Use With (Equipment) DR3900, DR6000, DR1900, DR2800, DR3800, DR5000
No. of Tests 25
Quantity 1 Pack
Range 0.015 to 2.0 and 0.02 to 2.5mg/L
Storage Requirements 2°C to 8°C
Test Method Indophenol blue
Type Ammonium Cuvette Test
compliance-icons
Product Identifier
  • Ammonium, Sample cuvette
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Sodium salicylate, Sodium hydroxide; caustic soda
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.