missing translation for 'onlineSavingsMsg'
Learn More

Hach Lange™ Cuvette Tests

Product Code. 10619741
Change view
Click to view available options
Type:
Ammonium Cuvette Test
Phosphate Ortho/Total Cuvette Test
Unit Size:
Each
Pack of 25
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
10619741 Phosphate Ortho/Total Cuvette Test Pack of 25
10594493 Ammonium Cuvette Test Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 10619741 Supplier Hach Lange™ Supplier No. LCK350
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

For use with water analysis, these exemplary cuvette tests have ISO 6878-1-1986 and DIN 38405 D11-4 certifications. They utilize the Phosphormolybdenum blue testing method for the DR3900, DR6000, DR1900, DR2800, DR3800, DR5000 instruments.

  • Phosphate (ortho/total) cuvette test 2.0 -20.0 mg/L PO4-P
    • ISO 6878-1-1986, DIN 38405 D11-4 certifications
    • 2 to 20 and 6 to 60mg/L range
    • For use with Water analysis
    • For use with DR3900, DR6000, DR1900, DR2800, DR3800, DR5000 instruments
    • 25 Tests
    • Shelf life of 24 Months
    • Phosphormolybdenum blue testing method
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO 6878-1-1986, DIN 38405 D11-4
For Use With (Application) Water analysis
For Use With (Equipment) DR3900, DR6000, DR1900, DR2800, DR3800, DR5000
No. of Tests 25
Quantity 1 Pack
Range 2 to 20 and 6 to 60mg/L
Storage Requirements 15°C to 25°C
Test Method Phosphormolybdenum blue
Type Phosphate Ortho/Total Cuvette Test
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Phosphate ortho/total cuvette test measuring range 2.0-20 mg/l PO4-P
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Reproductive toxicity Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H361-Suspected of damaging fertility or the unborn child.
  • H361d-Suspected of damaging the unborn child.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P390-Absorb spillage to prevent material damage.
Supplemental information
  • MIXTURE LIST-Contain: L-Ascorbic acid, Sodium metaborate tetrahydrate, Dextran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.