missing translation for 'onlineSavingsMsg'
Learn More

Globe Scientific Cuvettes for Mindray™ BS200

Product Code. 31922512 Shop All Globe Scientific Products
Change view
Click to view available options
Quantity:
1000/Cs.
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31922512 1000/Cs. Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31922512 Supplier Globe Scientific Supplier No. 5145
Estimated Shipment: 24-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed for use with Mindray™ BS200 chemistry analyzers. Globe Scientific Cuvettes for Mindray™ BS200 are produced from polystyrene (PS) and are offered individually wrapped in cases of 1000.

  • For use with Mindray™ BS200 chemistry analyzers
  • Cuvette segments are produced from polystyrene (PS)
  • Individually wrapped
  • Natural color

Specifications

Color Natural
Quantity 1000/Cs.
Material Polystyrene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.