missing translation for 'onlineSavingsMsg'
Learn More

CV-6209, TRC

Product Code. 30351660
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30351660 1 mg 1mg
30358585 10 mg 10mg
30360193 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30351660 Supplier Toronto Research Chemicals Supplier No. TRCC8560001MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100488-87-7
Molecular Formula C34 H60 N3 O6 . Cl
Molecular Weight (g/mol) 642.31
Synonym CV 6209
IUPAC Name [2-methoxy-3-(octadecylcarbamoyloxy)propyl] N-acetyl-N-[(1-ethylpyridin-1-ium-2-yl)methyl]carbamate;chloride
SMILES [Cl-].CCCCCCCCCCCCCCCCCCNC(=O)OCC(COC(=O)N(Cc1cccc[n+]1CC)C(=O)C)OC

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C34H59N3O6.ClH/c1-5-7-8-9-10-11-12-13-14-15-16-17-18-19-20-22-25-35-33(39)42-28-32(41-4)29-43-34(40)37(30(3)38)27-31-24-21-23-26-36(31)6-2;/h21,23-24,26,32H,5-20,22,25,27-29H2,1-4H3;1H
Formula Weight 641.4171
Chemical Name or Material CV-6209
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.