missing translation for 'onlineSavingsMsg'
Learn More

CX-5461 dihydrochloride, MedChemExpress

Product Code. 16435648
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16435648

Brand: Medchem Express HY13323A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CX-5461 dihydrochloride is a potent and orally bioavailable inhibitor of Pol I-mediated rRNA synthesis, with IC50s of 142 nM in HCT-116, 113 nM in A375, and 54 nM in MIA PaCa-2 cells, and shows little or no effect on Pol II (IC50 ≥25 μM).

CX-5461 dihydrochloride is a potent and orally bioavailable inhibitor of Pol I-mediated rRNA synthesis, with IC50s of 142 nM in HCT-116, 113 nM in A375, and 54 nM in MIA PaCa-2 cells, and shows little or no effect on Pol II (IC50 ≥25 μM).

Specifications

Chemical Name or Material CX-5461 dihydrochloride
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C27H29Cl2N7O2S
Quantity 10 mg
Solubility Information H2O : ≥ 25 mg/mL (42.62 mM)
SMILES CC1=NC=C(CNC(C2=C3N(C4=NC(N5CCN(CCC5)C)=CC=C4C2=O)C6=C(S3)C=CC=C6)=O)N=C1.[2 HCl]
Molecular Weight (g/mol) 586.54
Formula Weight 586.54
Percent Purity 98.07%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.