Learn More
CXCL16 Rat anti-Human, Clone: 256213, R&D Systems™
GREENER_CHOICE
Rat Monoclonal Antibody has been used in 1 publication
Brand:  R&D Systems MAB976-SP
Encompass
Product Code. 15720505
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
CXCL16 Monoclonal specifically detects CXCL16 in Human samples. It is validated for Western Blot, Flow Cytometry, Neutralization, CyTOF-ready.