missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL8/IL-8 Mouse anti-Human, Alexa Fluor 700, Clone: 6217, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC208N-100UG
This item is not returnable.
View return policy
Description
CXCL8/IL-8 Monoclonal specifically detects CXCL8/IL-8 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
| CXCL8/IL-8 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| P10145 | |
| IL8 | |
| E. coli-derived recombinant human CXCL8/IL-8 Ser28-Ser99 Accession # P10145 | |
| RUO | |
| 3576 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 6217 | |
| Alexa Fluor 700 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| 3-10C, AMCF-I, C-X-C motif chemokine 8, CXCL8, CXCL8SCYB8, Emoctakin, GCP1, GCP-1TSG-1, IL8, interleukin 8, K60, LAI, LECT, LUCT, LYNAP, MDNCF, MDNCFb-ENAP, member 8, MONAP, MONAPGCP1, NAF, NAP1, NAP-1NAP1, NCF, Neutrophil-activating protein 1, Protein 3-10C, T cell chemotactic factor, T-cell chemotactic factor, TCF, TSG1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human CXCL8/IL-8 in ELISAs and Western blots. In Western blots, this antibody shows 100% cross-reactivity with recombinant porcine CXCL8/IL-8 and no cross-reactivity with rrCXCL3/CINC-2 alpha . | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction