Learn More
GE Healthcare Cy5 Streptavidin
Product Code. 12974730 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
1mg
Product Code. Quantity unitSize
12974730 1 Ea. 1mg
1 options
Product Code. 12974730
Brand: GE Healthcare PA92005
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
  • Streptavidin conjugated to a range of fluorescent dyes for use in in situ hybridisation and flow cytometry.
  • High specificity reagents.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Ea.
Product Type Streptavidin-Fluor Conjugate
Signal Word
  • Danger
Hazard Category
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H317-May cause an allergic skin reaction.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501-Dispose of contents/container to .