missing translation for 'onlineSavingsMsg'
Learn More

Cyanamide, 50% w/w aq. soln., stab.

Product Code. 15439179
Change view
Click to view available options
Quantity:
100 g
250 g
1000 g
Unit Size:
1000g
100g
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15439179 250 g 250g
15429179 100 g 100g
15419179 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 15439179 Supplier Thermo Scientific Alfa Aesar Supplier No. L20448.30
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 420-04-2 | CH2N2 | 42.041 g/mol

It is widely used in the production of pharmaceuticals and other organic compounds. Cyanamide was used as nitrogen source in thermal transformation of metal oxide nanoparticles into nanocrystalline metal nitrides. It was used in the synthesis of mesoporous graphitic C3N4. Cyanamide in aqueous solution or in the form of its calcium salt is used as a fertilizer in agriculture. It is a potent inhibitor of liver aldehyde dehydrogenases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is widely used in the production of pharmaceuticals and other organic compounds. Cyanamide was used as nitrogen source in thermal transformation of metal oxide nanoparticles into nanocrystalline metal nitrides. It was used in the synthesis of mesoporous graphitic C3N4. Cyanamide in aqueous solution or in the form of its calcium salt is used as a fertilizer in agriculture. It is a potent inhibitor of liver aldehyde dehydrogenases.

Solubility
Soluble in water (85 g/100 ml (25°C). Soluble in organic solvents.

Notes
May form precipitate on storage
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 420-04-2
Molecular Formula CH2N2
Molecular Weight (g/mol) 42.041
MDL Number MFCD00007572
InChI Key XZMCDFZZKTWFGF-UHFFFAOYSA-N
Synonym hydrogen, carbimide, amidocyanogen, carbamonitrile, cyanoamine, cyanogenamide, cyanogen nitride, n-cyanoamine, alzogur, dormex
PubChem CID 9864
ChEBI CHEBI:16698
IUPAC Name cyanamide
SMILES C(#N)N

Specifications

Density 1.06
pH 4.2
Flash Point None
Odor Odorless
Linear Formula H2NCN
Refractive Index 1.395
Quantity 250 g
UN Number UN2922
Beilstein 1732569
Sensitivity Light sensitive; May form a precipitate
Merck Index 14,2684
Solubility Information Soluble in water (85g/100 ml (25°C). Soluble in organic solvents.
Formula Weight 42.04
Concentration or Composition (by Analyte or Components) 50% w/w aq. soln.
Chemical Name or Material Cyanamide, stabilized
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cyanamide, 50% w/w aqueous solution
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H351-Suspected of causing cancer.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Cyanamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.