missing translation for 'onlineSavingsMsg'
Learn More

WTW™ Cyanide Test Kits

Product Code. 11781073
Change view
Click to view available options
Range:
0.010 to 0.500 mg/L
Unit Size:
Pack of 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range Unit Size
11781073 0.010 to 0.500 mg/L Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11781073 Supplier WTW™ Supplier No. 250344
  • ONLINE EXCLUSIVE PRICE
    184.00 €
    176.00 € / Pack of 25
    Save 8.00 €
    4% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Test materials for the AQA of photometric routine and water analysis for multi-level result certainty of photoLab™, pHotoFlex™ and test kits

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance DIN 38 405 D13/ISO 6703
Volume (Metric) 25 mL
For Use With (Application) Measurement of Cyanide in wastewater from electroplating and metal processing works, drinking water
For Use With (Equipment) PhotoLab S6, PhotoLab S12, PhotoLab Spectral, Photoflex 0.01 - 0.30mg/L CN
Quantity 100 Pack
Range 0.010 to 0.500 mg/L
Type Test powder
compliance-icons
Product Identifier
  • Cyanide Cell Test (free and readily liberated cyanide) Method: photometric 0.010 - 0.500 mg/l CN
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
Supplemental information
  • MIXTURE LIST-Contain: troclosene sodium, dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.