missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cyanocobalamin, 96%

Product Code. 10627564 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
500mg
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10627564 5 g Glass bottle 5g
10420651 500 mg Glass bottle 500mg
10327291 1 g Glass bottle 1g
10713734 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10627564 Supplier Thermo Scientific Chemicals Supplier No. 405920050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 68-19-9 | C63H88CoN14O14P | 1355.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-19-9
Molecular Formula C63H88CoN14O14P
Molecular Weight (g/mol) 1355.38
MDL Number MFCD00151092
InChI Key AGVAZMGAQJOSFJ-WZHZPDAFSA-M
Synonym vitamin b12
PubChem CID 129893524
IUPAC Name cobalt(2+);[(2R,3S,4R,5S)-5-(5,6-dimethylbenzimidazol-1-yl)-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl] [(2R)-1-[3-[(1R,2R,3R,5Z,7S,10Z,12S,13S,15Z,17S,18S,19R)-2,13,18-tris(2-amino-2-oxoethyl)-7,12,17-tris(3-amino-3-oxopropyl)-3,5,8,8,13,15,18,19-octamethyl-
SMILES CC1=CC2=C(C=C1C)N(C=N2)C3C(C(C(O3)CO)OP(=O)(O)OC(C)CNC(=O)CCC4(C(C5C6(C(C(C(=C(C7=NC(=CC8=NC(=C(C4=N5)C)C(C8(C)C)CCC(=O)N)C(C7(C)CC(=O)N)CCC(=O)N)C)[N-]6)CCC(=O)N)(C)CC(=O)N)C)CC(=O)N)C)O.[C-]#N.[Co+2]

Specifications

Absorbance 3.15 to 3.45 (A361/A(547 to 559)), 1.7 to 1.9 (A361/A278)
Color Red
Loss on Drying 12% max. (20mg, 105°C, 2 hrs) (vacuum)
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Quantity 5 g
Merck Index 15, 10212
Solubility Information Solubility in water: soluble
Formula Weight 1355.38
Percent Purity 96%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Cyanocobalamin, 96%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.