missing translation for 'onlineSavingsMsg'
Learn More

Cyanocobalamin (Crystalline (Dark Red)), Fisher BioReagents

Product Code. 10533135
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10533135 1 g Amber Glass 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10533135 Supplier Fisher BioReagents Supplier No. BP8621
Quantity Price
1 72.77 € / 1g
10 66.33 € / 1g

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Cyanocobalamin is used as a supplement in cell culture and tissue culture media.
TRUSTED_SUSTAINABILITY

Specifications

CAS 68-19-9
Color Red
Assay Percent Range ≥98 %
Packaging Amber Glass
Molecular Formula C63H88CoN14O14P
MDL Number MFCD00151092
Quantity 1 g
Merck Index 15, 10212
Synonym vitamin b12
InChI Key AGVAZMGAQJOSFJ-WZHZPDAFSA-M
SMILES CC1=CC2=C(C=C1C)N(C=N2)C3C(C(C(O3)CO)OP(=O)(O)OC(C)CNC(=O)CCC4(C(C5C6(C(C(C(=C(C7=NC(=CC8=NC(=C(C4=N5)C)C(C8(C)C)CCC(=O)N)C(C7(C)CC(=O)N)CCC(=O)N)C)[N-]6)CCC(=O)N)(C)CC(=O)N)C)CC(=O)N)C)O.[C-]#N.[Co+2]
IUPAC Name cobalt(2+);[(2R,3S,4R,5S)-5-(5,6-dimethylbenzimidazol-1-yl)-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl] [(2R)-1-[3-[(1R,2R,3R,5Z,7S,10Z,12S,13S,15Z,17S,18S,19R)-2,13,18-tris(2-amino-2-oxoethyl)-7,12,17-tris(3-amino-3-oxopropyl)-3,5,8,8,13,15,18,19-octamethyl-
Molecular Weight (g/mol) 1356.396
PubChem CID 129893524
Formula Weight 1355.38
Percent Purity ≥98%
Physical Form Solid
Chemical Name or Material Cyanocobalamin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.