missing translation for 'onlineSavingsMsg'
Learn More

(Cyanomethyl)triphenylphosphonium chloride, 98+%

Product Code. 11494424
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11494424 100 g 100g
11384539 5 g 5g
11484424 25 g 25g
11404434 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11494424 Supplier Thermo Scientific Alfa Aesar Supplier No. A13096.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4336-70-3 | C20H17ClNP | 337.79 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4336-70-3
Molecular Formula C20H17ClNP
Molecular Weight (g/mol) 337.79
MDL Number MFCD00031672
InChI Key ARPLQAMUUDIHIT-UHFFFAOYSA-M
Synonym cyanomethyl triphenylphosphonium chloride, phosphonium, cyanomethyl triphenyl-, chloride, cyanomethyl triphenylphosphanium chloride, cyanomethyl triphenyl phosphanium chloride, cyanomethyltriphenylphosphonium chloride, acmc-1ctgb, wln: nc1pr&r&r &g, cyanomethyl triphenylphosphoniumchloride, cyanomethyl triphenyl phosphonium chloride
PubChem CID 197040
IUPAC Name cyanomethyl(triphenyl)phosphanium;chloride
SMILES [Cl-].N#CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point ∼270°C (decomposition)
Quantity 100 g
UN Number UN3439
Beilstein 6222047
Sensitivity Hygroscopic
Formula Weight 337.79
Percent Purity ≥98%
Chemical Name or Material (Cyanomethyl)triphenylphosphonium chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.