missing translation for 'onlineSavingsMsg'
Learn More

Cyantraniliprole, MedChemExpress

Product Code. 16488057
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488057 5 mg 5mg
16471537 50 mg 50mg
16455976 100 mg 100mg
16415348 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16488057 Supplier Medchem Express Supplier No. HY12779
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Cyantraniliprole(HGW-86) is an insecticide of the ryanoid class; has activity against pests such as Diaphorina citri that have developed resistance to other classes insecticides.

Cyantraniliprole(HGW-86) is an insecticide of the ryanoid class

Specifications

Chemical Name or Material Cyantraniliprole
CAS 736994-63-1
Color White
Purity Grade Notes Research
Molecular Formula C19H14BrClN6O2
Quantity 5 mg
Synonym HGW-86
Solubility Information DMSO : 50 mg/mL (105.55 mM; Need ultrasonic)
SMILES O=C(C1=CC(Br)=NN1C2=NC=CC=C2Cl)NC3=C(C(NC)=O)C=C(C#N)C=C3C
Molecular Weight (g/mol) 473.71
Formula Weight 473.71
Percent Purity 99.83%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.