missing translation for 'onlineSavingsMsg'
Learn More

Cyanuric chloride, 98%

Product Code. 11362156
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11362156 250 g 250g
11352156 50 g 50g
11372156 1000 g 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11362156 Supplier Thermo Scientific Alfa Aesar Supplier No. L03442.30
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 108-77-0 | C3Cl3N3 | 184.40 g/mol

Reagent for detection of glycine in presence of other amino acids.Cyanuric chloride is a reagent used for the conversion of alcohols and carboxylic acids to alkyl chlorides and acid chlorides respectively. It acts as an intermediate to prepare dyestuffs, optical brighteners, tanning agents, UV-absorbing agent, softening agents and pharmaceuticals. It is also used as a precursor to dyes and crosslinking agents due to the presence of reactive chlorine atoms. It is used as a dehydrating agent as well as reacting with dimethylformamide to give Gold’s reagent, which is a versatile source of aminoalkylations and a precursor to heterocycles. Further, it is considered as an alternative to oxalyl chloride in the Swern oxidation. In addition to this, it is involved in the synthesis of adenosine receptor ligand.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent for detection of glycine in presence of other amino acids.Cyanuric chloride is a reagent used for the conversion of alcohols and carboxylic acids to alkyl chlorides and acid chlorides respectively. It acts as an intermediate to prepare dyestuffs, optical brighteners, tanning agents, UV-absorbing agent, softening agents and pharmaceuticals. It is also used as a precursor to dyes and crosslinking agents due to the presence of reactive chlorine atoms. It is used as a dehydrating agent as well as reacting with dimethylformamide to give Gold’s reagent, which is a versatile source of aminoalkylations and a precursor to heterocycles. Further, it is considered as an alternative to oxalyl chloride in the Swern oxidation. In addition to this, it is involved in the synthesis of adenosine receptor ligand.

Solubility
Soluble in acetone, diethyl ether, dioxane, chloroform, carbon tetrachloride and ketones. Slightly soluble in methanol. Insoluble in cold water.

Notes
Air and moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents, strong acids, alcohols, dimethylformamide, amines and dimethyl sulfoxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-77-0
Molecular Formula C3Cl3N3
Molecular Weight (g/mol) 184.40
MDL Number MFCD00006046
InChI Key MGNCLNQXLYJVJD-UHFFFAOYSA-N
Synonym cyanuric chloride, cyanuryl chloride, cyanuric acid chloride, s-triazine trichloride, trichlorocyanidine, cyanuric trichloride, tricyanogen chloride, sym-trichlorotriazine, cyanur chloride, trichloro-s-triazine
PubChem CID 7954
ChEBI CHEBI:58964
SMILES ClC1=NC(Cl)=NC(Cl)=N1

Spezifikation

Melting Point 145°C to 149°C
Density 1.92
Boiling Point 192°C to 194°C
Flash Point >190°C (374°F)
Quantity 250 g
UN Number UN2670
Beilstein 124246
Sensitivity Moisture sensitive
Solubility Information Soluble in acetone,diethyl ether,dioxane,chloroform,carbon tetrachloride and ketones. Slightly soluble in methanol. Insoluble in cold water.
IUPAC Name 2,4,6-trichloro-1,3,5-triazine
Formula Weight 184.41
Percent Purity 0.98
Chemical Name or Material Cyanuric chloride
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.