missing translation for 'onlineSavingsMsg'
Learn More

Cyclamic Acid Sodium Salt, TRC

Product Code. 30138576
Change view
Click to view available options
Quantity:
100 g
250 g
Unit Size:
100g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138576 250 g 250g
17943745 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138576 Supplier Toronto Research Chemicals Supplier No. TRCC987635250G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139-05-9
Molecular Formula C6 H12 N O3 S . Na
Molecular Weight (g/mol) 201.22
Synonym Cyclohexanesulfamic acid, monosodium salt (8CI), Sulfamic acid, cyclohexyl-, monosodium salt (9CI), Acofarinas, Assugrin, Asugryn, Azucrona, Cyclamate sodium, Cyclohexylsulfamate sodium, Cyclohexylsulfamic acid sodium salt, Dulzor-Etas, Duplo-Sacarin, Hachi-Sugar, Ibiosuc, N-Cyclohexylsulfamic acid sodium salt, Natreen, Nutter, Sodium cyclamate, Sodium cyclohexanesulfamate, Sodium cyclohexylaminesulfonate, Sodium cyclohexylsulfamate, Sodium sucaryl, Sucaryl sodium, Sucrum 7, Suessette, Suestamin, Sugarin, Sugaron
IUPAC Name sodium;N-cyclohexylsulfamate
SMILES [Na+].[O-]S(=O)(=O)NC1CCCCC1

Specifications

Quantity 250 g
InChI Formula InChI=1S/C6H13NO3S.Na/c8-11(9,10)7-6-4-2-1-3-5-6;/h6-7H,1-5H2,(H,8,9,10);/q;+1/p-1
Formula Weight 201.0436
Percent Purity >95%
Chemical Name or Material Sodium cyclamate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.