Learn More
Invitrogen™ Cyclic GMP Competitive ELISA Kit
Description
The cyclic GMP Competitive ELISA quantitates cGMP in cell or tissue lysates. The assay will exclusively recognize both natural and recombinant cyclic GMP. Principle of the method The cyclic GMP solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is inversely proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Specifications
Specifications
| Assay Range | 0.8-5000 pmol/mL |
| Assay Sensitivity | 0.059 pmol/mL |
| Description | cGMP Competitive ELISA Kit |
| Conjugate | AP |
| Immunoassay Kit Format | Colorimetric ELISA (p-NPP substrate) with Plates |
| Product Type | ELISA Kit |
| Sample Type | Cell Lysates, Supernatant, Tissue Homogenates |
| Specificity | No significant cross-reaction with GMP, GTP, cAMP, AMP, ATP, cUMP, or CTP |
| Sufficient For | 96 microplate wells |
| Target Species | Human |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.