missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Cyclic Pifithrin-α hydrobromide

Product Code. 18085402
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18085402 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18085402 Supplier R&D Systems™ Supplier No. 3843/10
  • ONLINE EXCLUSIVE PRICE
    273.17 €
    237.30 € / 10mg
    Save 35.87 €
    13% Off
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. p53 inhibitor

Cyclic Pifithrin-α hydrobromide is a cyclic analog of pifithrin-α (Cat. No. 1267), a small molecule inhibitor of p53. Prevents dexamethasone -induced cell death in murine thymocytes (EC50 = 2.01 μM). Sensitizes p53-deficient tumors to radiotherapy and chemotherapy; increases apoptosis in target cells when used in combination with antimicrotubule agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 511296-88-1
Molecular Formula C16H17BrN2S

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 349.29
Purity >98%
Molecular Weight (g/mol) 349.29
Chemical Name or Material Cyclic Pifithrin-α hydrobromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.