missing translation for 'onlineSavingsMsg'
Learn More

Cyclobutanecarboxylic acid chloride, 99%

Product Code. 10102620 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10102620 10 g Glass bottle 10g
10487573 1 g Glass bottle 1g
10423775 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10102620 Supplier Thermo Scientific Acros Supplier No. 110910100
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5006-22-4 | C5H7ClO | 118.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5006-22-4
Molecular Formula C5H7ClO
Molecular Weight (g/mol) 118.56
MDL Number MFCD00001319
InChI Key JFWMYCVMQSLLOO-UHFFFAOYSA-N
Synonym cyclobutanecarboxylic acid chloride, cyclobutanecarbonylchloride, cyclobutane carbonyl chloride, cyclobutane-1-carbonyl chloride, cyclobutancarbonylchlorid, cyclobutylcarbonylchloride, cyclobutancarbonyl chloride, cyclobutyl carbonylchloride, cyclobutylcarbonyl chloride, acmc-1aqy1
PubChem CID 78705
IUPAC Name cyclobutanecarbonyl chloride
SMILES C1CC(C1)C(=O)Cl

Specifications

Color Colorless
Boiling Point 145.0°C
Flash Point 37°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula C4H7COCl
Refractive Index 1.4545 to 1.4565
Quantity 10 g
Beilstein 09, 5
Formula Weight 118.56
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Cyclobutanecarboxylic acid chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cyclobutanecarboxylic acid chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.