missing translation for 'onlineSavingsMsg'
Learn More

Cyclobutanecarboxylic Acid, TRC

Product Code. 30350109
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity unitSize
30350109 1 g 1g
1 options
This item is not returnable. View return policy

Product Code. 30350109

Brand: Toronto Research Chemicals TRCC9881651G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cyclobutanecarboxylic Acid
CAS 3721-95-7
Molecular Formula C5 H8 O2
Quantity 1 g
Synonym Cyclobutanecarboxylic Acid, Cyclobutane-1-carboxylic acid, Cyclobutylcarboxylic Acid, NSC 4535
InChI Formula InChI=1S/C5H8O2/c6-5(7)4-2-1-3-4/h4H,1-3H2,(H,6,7)
SMILES OC(=O)C1CCC1
IUPAC Name cyclobutanecarboxylic acid
Molecular Weight (g/mol) 100.12
Formula Weight 100.0524
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.