missing translation for 'onlineSavingsMsg'
Learn More

Cyclobutylamine, 98%

Product Code. 11425463
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11425463 25 g 25g
11405463 1 g 1g
11415463 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11425463 Supplier Thermo Scientific Alfa Aesar Supplier No. A13423.14
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2516-34-9 | C4H9N | 71.123 g/mol

Substrate employed in a microwave-assisted synthesis of 7-azaindoles from dihalopyridines. Also it is a reactant in the preparation of imidazo[1,2-b]pyridazine derivatives as selective and orally available Mps1 (TTK) kinase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Substrate employed in a microwave-assisted synthesis of 7-azaindoles from dihalopyridines. Also it is a reactant in the preparation of imidazo[1,2-b]pyridazine derivatives as selective and orally available Mps1 (TTK) kinase inhibitors.

Solubility
Slightly soluble in water.

Notes
Hygroscopic, store way form water/moisture. Store in cool, dry place in tightly closed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2516-34-9
Molecular Formula C4H9N
Molecular Weight (g/mol) 71.123
MDL Number MFCD00001328
InChI Key KZZKOVLJUKWSKX-UHFFFAOYSA-N
Synonym cyclobutylamine, aminocyclobutane, cyclobutyl amine, cyclobutylamin, cyclobutaneamine, 1-amino cyclobutane, pubchem11047, acmc-209ghn, cyclobutylammonium chloride
PubChem CID 75645
IUPAC Name cyclobutanamine
SMILES C1CC(C1)N

Specifications

Density 0.836
Boiling Point 82°C to 83°C
Flash Point −4°C (24°F)
Refractive Index 1.4376
Quantity 25 g
UN Number UN2733
Beilstein 2069297
Sensitivity Air sensitive; Hygroscopic
Solubility Information Slightly soluble in water.
Formula Weight 71.12
Percent Purity 98%
Chemical Name or Material Cyclobutylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.