missing translation for 'onlineSavingsMsg'
Learn More

Cyclocytidine hydrochloride, 98+%, Thermo Scientific Chemicals

Product Code. 15416009
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15416009 1 g 1g
15426009 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15416009 Supplier Thermo Scientific Alfa Aesar Supplier No. J63845.03
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Anti-tumor agent

Anti-tumor agent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10212-25-6
Molecular Formula C9H12ClN3O4
Molecular Weight (g/mol) 261.66
MDL Number MFCD00012636
InChI Key KZOWNALBTMILAP-JBMRGDGGSA-N
Synonym cyclocytidine hydrochloride 1g
PubChem CID 74764394
ChEBI CHEBI:74843
IUPAC Name (2R,3R,3aS,9aR)-2-(hydroxymethyl)-6-imino-2,3,3a,9a-tetrahydrofuro[1,2][1,3]oxazolo[3,4-a]pyrimidin-3-ol;hydron;chloride
SMILES Cl.OC[C@H]1O[C@@H]2[C@@H](OC3=NC(=N)C=CN23)[C@@H]1O

Specifications

Melting Point 248°C to 250°C
Color White
Quantity 1 g
Merck Index 14,629
Formula Weight 261.66
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Cyclocytidine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.