missing translation for 'onlineSavingsMsg'
Learn More

β-Cyclodextrin, >98%, MP Biomedicals™

Product Code. 11494502 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11494502 5 g 5g
11404512 25 g 25g
11414512 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11494502 Supplier MP Biomedicals Supplier No. 0219005305
 Restricted Item
Estimated Shipment: 14-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 7585-39-9
Molecular Formula C42H70O35
Molecular Weight (g/mol) 1134.99
MDL Number MFCD00078139
InChI Key WHGYBXFWUBPSRW-UHFFFAOYNA-N
Synonym cycloheptaamylose, cycloheptapentylose, .beta.-cyclodextrin, caraway, cyclo-hepta-amylose, cycloheptamaltose, .beta.-dextrin, beta-cycloamylose, beta-cyclodextrin, .beta.-cycloamylose
PubChem CID 131707246
IUPAC Name 5,10,15,20,25,30,35-heptakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontane-36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecol
SMILES OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC1C(O)C2O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O

Specifications

Color White
Assay Percent Range >98%
Quantity 5 g
Formula Weight 1134.9
Physical Form Powder
Chemical Name or Material β-Cyclodextrin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.