missing translation for 'onlineSavingsMsg'
Learn More

beta-Cyclodextrin hydrate, 99%

Product Code. 10665272 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10665272 5 g Glass bottle 5g
10308560 25 g Glass bottle 25g
10532251 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10665272 Supplier Thermo Scientific Acros Supplier No. 227280050
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 68168-23-0 | C42H70O35 | 1134.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68168-23-0
Molecular Formula C42H70O35
Molecular Weight (g/mol) 1134.99
MDL Number MFCD00150811
InChI Key WHGYBXFWUBPSRW-UHFFFAOYSA-N
Synonym beta-cyclodextrin hydrate, schardinger, a-dextrin hydrate, beta-cyclodextrin hydrate, puriss hplc
PubChem CID 53486154
IUPAC Name 5,10,15,20,25,30,35-heptakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontane-36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecol
SMILES OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC1C(O)C2O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 298°C to 300°C
Color White
pH 5.0 to 8.0 (1% aq. soln.)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 5 g
Fieser 06,151; 08,133; 09,129; 12,151; 15,107
Merck Index 14, 2718
Specific Rotation + 161
Solubility Information (1% in water) Clear colorless
Residual Solvents 5ppm toluene max.
Formula Weight 1135.01
Percent Purity 99%
Related Substances 1% max. Alpha-and gamma-cyclodextrin (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material β-Cyclodextrin hydrate, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.