missing translation for 'onlineSavingsMsg'
Learn More

gamma-Cyclodextrin hydrate, 99%

Product Code. 10144810 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10144810 500 mg Glass bottle 500mg
10753041 1 g Glass bottle 1g
10247123 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10144810 Supplier Thermo Scientific Acros Supplier No. 229905000
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 91464-90-3 | C48H80O40 | 1297.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91464-90-3
Molecular Formula C48H80O40
Molecular Weight (g/mol) 1297.13
MDL Number MFCD00149574
InChI Key GDSRMADSINPKSL-UHFFFAOYNA-N
Synonym gamma-cyclodextrin hydrate, 1297.12 anhydrous, gamma-cyclodextrin hydrate, vetec tm reagent grade
PubChem CID 71311509
IUPAC Name 5,10,15,20,25,30,35,40-octakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34,37,39-hexadecaoxanonacyclo[36.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹.2³³,³⁶]hexapentacontane-41,42,43,44,45,46,47,48,49,50,51,52,53,54,55,56-hexadecol
SMILES OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC9C(CO)OC(OC1C(O)C2O)C(O)C9O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O

Specifications

Melting Point 267.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 500 mg
Merck Index 14, 2718
Specific Rotation + 177.00
Solubility Information Solubility in water: 22.3 g/100 mL (25°C).
Formula Weight 1297.15
Percent Purity 99%
Physical Form Powder
Chemical Name or Material γ-Cyclodextrin hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.