missing translation for 'onlineSavingsMsg'
Learn More

α-Cyclodextrin, MedChemExpress

Product Code. 16452349
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16452349 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16452349 Supplier Medchem Express Supplier No. HYB1513
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress α-Cyclodextrin is a multifunctional, soluble dietary fiber marketed for use as a fiber ingredient.

α-Cyclodextrin is a multifunctional, soluble dietary fiber marketed for use as a fiber ingredient.

Specifications

Chemical Name or Material α-Cyclodextrin
CAS 10016-20-3
Color White
Purity Grade Notes Research
Molecular Formula C36H60O30
Quantity 1 g
Solubility Information DMSO : 110 mg/mL (113.07 mM; Need ultrasonic) ∣H2O : 50 mg/mL (51.40 mM; Need ultrasonic)
SMILES OC[C@@H](O[C@@](O[C@]([C@@H]1O)([H])[C@H](O[C@@](O[C@]([C@@H]2O)([H])[C@H](O[C@@](O[C@@]3([H])[C@@H]4O)([H])[C@@H]2O)CO)([H])[C@@H]1O)CO)([H])[C@@H]5O)[C@](O[C@](O[C@H](CO)[C@](O[C@](O[C@H](CO)[C@](O[C@](O[C@@H]3CO)([H])[C@@H]4O)([H])[C@@H]6O)([H])[C@@H]6O)([H])[C@@H]7O)([H])[C@@H]7O)([H])[C@@H]5O
Molecular Weight (g/mol) 972.84
Formula Weight 972.84
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.