missing translation for 'onlineSavingsMsg'
Learn More

β-Cyclodextrin, MP Biomedicals™

Product Code. 11484502 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11484502 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11484502 Supplier MP Biomedicals Supplier No. 0219005301
 Restricted Item
Estimated Shipment: 14-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 7585-39-9
Density 1.624g/cm3 (L)
Molecular Formula C42H70O35
MDL Number MFCD00078139
Quantity 1 g
Synonym cycloheptaamylose, cycloheptapentylose, .beta.-cyclodextrin, caraway, cyclo-hepta-amylose, cycloheptamaltose, .beta.-dextrin, beta-cycloamylose, beta-cyclodextrin, .beta.-cycloamylose
InChI Key WHGYBXFWUBPSRW-UHFFFAOYNA-N
SMILES OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC1C(O)C2O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O
IUPAC Name 5,10,15,20,25,30,35-heptakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontane-36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecol
Molecular Weight (g/mol) 1134.99
PubChem CID 131707246
Formula Weight 1134.9
Physical Form Solid
Chemical Name or Material β-Cyclodextrin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.