missing translation for 'onlineSavingsMsg'
Learn More

α-Cyclodextrin (White Powder), Fisher BioReagents

Product Code. 10695215
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Amber Glass
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10695215 5 g Amber Glass 5g
10378024 25 g Amber Glass 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10695215 Supplier Fisher BioReagents Supplier No. BP25015

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
  • A cyclic polysaccharide comprised of 6 glucose units that improves the solubility of hydrophobic molecules
  • Improves the solubility of nonpolar molecules such as fatty acids, lipids, and cholesterol in aqueous media
  • Also used for selective precipitation of enantiomeric, positional or structural isomers in chromatrographic separations and purification methods
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 10016-20-3
Molecular Formula C36H60O30
Molecular Weight (g/mol) 972.846
InChI Key HFHDHCJBZVLPGP-UHFFFAOYSA-N
Synonym cyclohexaamylose, cyclomaltohexose, .alpha.-cyclodextrin, schardinger alpha-dextrin, .alpha.-dextrin, .alpha.-cycloamylose, bdbm4, dextrin, .alpha.-cyclo, .alpha.-schardinger dextrin
PubChem CID 320760
SMILES C(C1C2C(C(C(O1)OC3C(OC(C(C3O)O)OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)OC6C(OC(C(C6O)O)OC7C(OC(O2)C(C7O)O)CO)CO)CO)CO)CO)O)O)O

Spezifikation

Melting Point 278°C
Color White
Loss on Drying 13% max.
Packaging Amber Glass
Quantity 5 g
Merck Index 15, 2710
Specific Rotation 143 to 153° (+ or -)
Formula Weight 972.9
Physical Form Powder or Solid
Chemical Name or Material α-Cyclodextrin
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.