missing translation for 'onlineSavingsMsg'
Learn More

Cyclogalegenin, MedChemExpress

Product Code. 16448978
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16448978 10 mg 10mg
16424837 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16448978 Supplier Medchem Express Supplier No. HYN0424

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cyclogalegenin (Cyclogalegigenin) is a isoprenoid found in Astragalus galegiformis. Cyclogalegenin is the enantiomer of Cycloastragenol.

Cyclogalegenin (Cyclogalegigenin) is a isoprenoid found in Astragalus galegiformis. Cyclogalegenin is the enantiomer of Cycloastragenol.

Specifications

Chemical Name or Material Cyclogalegenin
CAS 84605-18-5
Color White
Purity Grade Notes Research
Molecular Formula C30H50O5
Quantity 10 mg
Synonym Cyclogalegigenin
Solubility Information DMSO : 100 mg/mL (203.78 mM; Need ultrasonic)
SMILES O[C@H]1CC[C@@]2(C3)[C@]([C@@H](O)C[C@]4([H])[C@@]23CC[C@@]5(C)[C@@]4(C)C[C@H](O)[C@]5([H])[C@@]6(C)O[C@@H](C(C)(C)O)CC6)([H])C1(C)C
Molecular Weight (g/mol) 490.72
Formula Weight 490.72
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.