missing translation for 'onlineSavingsMsg'
Learn More

Cycloheptane, 99%

Product Code. 11399789
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11399789

Brand: Thermo Scientific Alfa Aesar B22902.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 291-64-5 | C7H14 | 98.19 g/mol

Cycloheptane is used as a nonpolar solvent for the chemical industry and as an intermediate in the manufacture of chemicals and pharmaceutical drugs. It can be used to get fluorocycloheptane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cycloheptane is used as a nonpolar solvent for the chemical industry and as an intermediate in the manufacture of chemicals and pharmaceutical drugs. It can be used to get fluorocycloheptane.

Solubility
Soluble in water 30 mg @ 20°C.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 291-64-5
Molecular Formula C7H14
Molecular Weight (g/mol) 98.19
MDL Number MFCD00004147
InChI Key DMEGYFMYUHOHGS-UHFFFAOYSA-N
Synonym heptamethylene, unii-vtz53p34ja, vtz53p34ja, cycloheptan, hsdb 59, cycloheptane, un2241 flammable liquid
PubChem CID 9265
IUPAC Name cycloheptane
SMILES C1CCCCCC1

Specifications

Density 0.811
Boiling Point 118°C to 119°C
Flash Point 6°C (43°F)
Refractive Index 1.445
Quantity 5 g
UN Number UN1206
Solubility Information Soluble in water 30 mg @ 20°C.
Formula Weight 98.19
Percent Purity 99%
Chemical Name or Material Cycloheptane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.