missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexane, 99.5%, for analysis

Product Code. 10429650 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
25 L
Packaging:
Glass Bottle
Plastic Drum
Unit Size:
1 L
2.5 L
25 L
5 L
500mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10429650 1 L Glass Bottle 1 L
10205950 500 mL Glass Bottle 500mL
10439650 2.5 L Glass Bottle 2.5 L
10536993 5 L Plastic Drum 5 L
10777773 25 L Plastic Drum 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10429650 Supplier Thermo Scientific Acros Supplier No. 176810010
  • ONLINE EXCLUSIVE PRICE
    75.85 €
    73.60 € / 1 L
    Save 2.25 €
    2% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 110-82-7 | C6H12 | 84.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-82-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00003814
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
Synonym hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
PubChem CID 8078
ChEBI CHEBI:29005
IUPAC Name cyclohexane
SMILES C1CCCCC1

Specifications

Melting Point 6.5°C
Density 0.7700g/mL
Quantity 1 L
Flash Point −18°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.4250 to 1.4270
Residue after Evaporation 0.005% max.
Assay Percent Range 99.5% min. (GC)
Specific Gravity 0.77
Linear Formula C6H12
Viscosity 0.94 mPa.s (20°C)
Water 0.05% max.
Beilstein 05, 20
Merck Index 15, 2716
Boiling Point 81.0°C
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol (57 g/100 mL at 20°C), ethanol, , diethyl ether, acetone, benzene, ccl4.
Formula Weight 84.15
Percent Purity 99.5%
Grade Analytical
Chemical Name or Material Cyclohexane
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cyclohexane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.