missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexane, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™

Product Code. 15244228 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15244228 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15244228 Supplier Thermo Scientific Acros Supplier No. 364665000
  • ONLINE EXCLUSIVE PRICE
    68.00 €
    62.50 € / 500mL
    Save 5.50 €
    8% Off
View all promotions
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 110-82-7 | C6H12 | 84.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 6.5°C
CAS 110-82-7
Density 0.7700g/mL
Quantity 500 mL
Flash Point −18°C
Refractive Index 1.4250 to 1.4270
Specific Gravity 0.77
Molecular Formula C6H12
Linear Formula C6H12
MDL Number MFCD00003814
Viscosity 0.94 mPa.s (20°C)
Water 0.005% max. (Coulometric)
Synonym hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
Boiling Point 81.0°C
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol (57 g/100 mL at 20°C), ethanol, , diethyl ether, acetone, benzene, ccl4.
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
SMILES C1CCCCC1
IUPAC Name cyclohexane
Molecular Weight (g/mol) 84.16
PubChem CID 8078
ChEBI CHEBI:29005
Formula Weight 84.15
Percent Purity 99.5%
Grade Extra Dry over Molecular Sieve
Chemical Name or Material Cyclohexane
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cyclohexane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.