missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexane, BAKER ANALYZED, J.T. Baker™

Product Code. 10201201 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Glass
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10201201 2.5 L Glass 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10201201 Supplier JT Baker Supplier No. 8026.2500
  • ONLINE EXCLUSIVE PRICE
    195.00 €
    177.20 € / 2.5 L
    Save 17.80 €
    9% Off
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High quality grade chemical that exceeds ACS specifications for laboratory and specialized industrial use.

  • Exceeds the requirements set forth by the American Chemical Society (ACS)
  • Label provides an actual Lot Analysis as a guarantee of purity and consistency
  • Analytical methods used for these determinations are based on standard test procedures described in technical publications
  • Improvements developed by J.T. Baker are employed to achieve a higher degree of reliability

Specifications

Melting Point 6.4°C
CAS 110-82-7
Color Colorless
Density 0.78g/cm3 at 20°C
Quantity 2.5 L
Explosion Limit 1.2 - 8.3 Vol%
Flash Point −18°C
Ignition Point 260°C
Odor Sweet
Packaging Glass
Physical Form Fluid
Residue after Evaporation Max. 0.002%
Molecular Formula C6H12
Linear Formula CH2(CH2)4CH2
MDL Number MFCD00003814
Viscosity 0.94 mPas at 20°C
Water Max. 0.02%, 0.05g/L
Synonym hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
Boiling Point 81°C
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
SMILES C1CCCCC1
IUPAC Name cyclohexane
Molecular Weight (g/mol) 84.16
PubChem CID 8078
ChEBI CHEBI:29005
Formula Weight 84.16g/mol
Percent Purity ≥99.0%
Grade Exceeds ACS specifications
Chemical Name or Material Cyclohexane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.