missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexane, BAKER ANALYZED Pesticide Reagent, J.T. Baker™

Product Code. 15272158 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Glass
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15272158 2.5 L Glass 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15272158 Supplier JT Baker Supplier No. 5278.2500
  • ONLINE EXCLUSIVE PRICE
    269.00 €
    242.77 € / 2.5 L
    Save 26.23 €
    9% Off
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specially developed for use in evaluation of pesticide and insecticide residues in food, feed, water or soil samples. Cyclohexane, BAKER ANALYZED Pesticide Reagent, J.T. Baker™ is suitable for all analysis of common pesticides.

J.T. Baker BAKER ANALYZED Pesticide Reagent Solvents:

  • Suitable for analysis of most common pesticides, such as 2-chlorobiphenyl , Heptachlor, Aldrin, Parathion, Dieldrin, Endrin, DDT, Dioxine, etc.
  • Specified to the required low residue levels (5 ppm) for use in research and quality control laboratories
  • Ideal for gas chromatography applications, testing for ECD and/or FID sensitive impurities, organic residues, and residue after evaporation

Specifications

Melting Point 6.4°C
CAS 110-82-7
Color Colorless
Acidity Max. 0.0005meq/g
Density 0.78g/cm3 at 20°C
Quantity 2.5 L
Explosion Limit 1.2 - 8.3 Vol %
Flash Point −18°C
Ignition Point 260°C
Odor Sweetish
Packaging Glass
Physical Form Liquid
Residue after Evaporation Max. 5 ppm
Molecular Formula C6H12
MDL Number MFCD00003814
Viscosity 0.94 mPas at 20°C
Water Max. 0.02%
Synonym hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
Boiling Point 81°C
Solubility Information 0.05 g/L in water
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
SMILES C1CCCCC1
IUPAC Name cyclohexane
Molecular Weight (g/mol) 84.16
PubChem CID 8078
ChEBI CHEBI:29005
Formula Weight 84.16g/mol
Percent Purity Min. 99.5%
Grade Pesticide
Chemical Name or Material Cyclohexane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.