missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexane, BAKER, J.T. Baker™

Product Code. 10721932 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10721932 1 L 1 L
10383311 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10721932 Supplier JT Baker Supplier No. 8104.1000
  • ONLINE EXCLUSIVE PRICE
    73.49 €
    55.79 € / 1 L
    Save 17.70 €
    24% Off
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For laboratory use, for organic synthesis, and for routine research.

A grade of chemicals for routine research usually controlled by physical criteria and often by assay (by GC).

Chemical Identifiers

CAS 110-82-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00003814
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
Synonym hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
PubChem CID 8078
ChEBI CHEBI:29005
IUPAC Name cyclohexane
SMILES C1CCCCC1

Specifications

Melting Point 6.4°C
Color Colorless
Density 0.78g/cm3 at 20°C
Quantity 1 L
Explosion Limit 1.2 - 8.3 Vol %
Flash Point −18°C
Ignition Point 260°C
Odor Sweetish
Packaging Glass
Physical Form Liquid
Residue after Evaporation Max. 0.005%
Linear Formula CH2(CH2)4CH2
Viscosity 0.94 mPas at 20°C
Water Max. 0.02%
Boiling Point 81°C
Solubility Information 0.05g/L in water
Formula Weight 84.16g/mol
Percent Purity ≥99%
Grade BAKER
Chemical Name or Material Cyclohexane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.