missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexane, CHROMASOLV™, for HPLC, ≥99.7%, Solstice

Product Code. 15644540
Change view
Click to view available options
Quantity:
100 mL
1 L
2 L
2.5 L
4 x 2.5 L
6 x 1 L
Packaging:
Glass Bottle
Unit Size:
1 L
100mL
2 L
2.5 L
Pack of 4
Pack of 6
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15644540 6 x 1 L Glass Bottle Pack of 6
15694530 100 mL Glass Bottle 100mL
15604540 1 L Glass Bottle 1 L
15624540 2 L Glass Bottle 2 L
15614540 2.5 L Glass Bottle 2.5 L
15634540 4 x 2.5 L Glass Bottle Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15644540 Supplier Honeywell-Chromasolv Supplier No. 348556X1L
  • ONLINE EXCLUSIVE PRICE
    690.00 €
    565.20 € / Pack of 6
    Save 124.80 €
    18% Off
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CHROMASOLV™, for HPLC,≥99.7%

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-82-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00003814
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
Synonym hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
PubChem CID 8078
ChEBI CHEBI:29005
IUPAC Name cyclohexane
SMILES C1CCCCC1

Specifications

Melting Point 4°C to 7°C (literature)
Absorbance max. 0.10 at 235 nm, max. 0.20 at 230 nm, max. 0.50 at 220 nm, max. 1.00 at 210 nm, max. 0.01 at 400 nm, max. 0.01 at 300 nm, max. 0.01 at 255 nm, max. 0.03 at 250 nm, max. 0.08 at 240 nm
CAS Max % ≤100.0000%
Density 0.779 g/mL at 25 °C(lit.); 0.780 g/mL at 25 °C
Quantity 6 x 1 L
Flash Point −18°C
Packaging Glass Bottle
Linear Formula C6H12
UN Number UN1145
Beilstein 1900225
Boiling Point 81°C
Percent Purity ≥99.7%
Grade HPLC
Chemical Name or Material Cyclohexane, CHROMASOLV™, for HPLC, ≥99.7%
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cyclohexane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.