missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexane-d{12}, 99.5%(Isotopic)

Product Code. 11399947
Change view
Click to view available options
Quantity:
10 Ea.
2 Ea.
Unit Size:
Pack of 10
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11399947 2 Ea. Pack of 2
11309957 10 Ea. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11399947 Supplier Thermo Scientific Alfa Aesar Supplier No. 042271.SA
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 1735-17-7 | C6H12 | 96.24 g/mol

It is a common solvent used in NMR spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1735-17-7
Molecular Formula C6H12
Molecular Weight (g/mol) 96.24
MDL Number MFCD00044212
InChI Key XDTMQSROBMDMFD-LBTWDOQPSA-N
Synonym cyclohexane-d12, 2h12 cyclohexane, cyclohexane d, cyclohexane, d12, 2 h?? cyclohexane, cyclohexane-d12, ≥99.6 atom % d, cyclohexane #, cyclohexane-d12, 99.5 atom % d
PubChem CID 123129
SMILES [2H]C1([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C1([2H])[2H]

Specifications

Physical Form Liquid
Density 0.89 g/mL
Flash Point -18°C (-1°F)
Refractive Index 1.421
Quantity 2 Ea.
Melting Point 6.5°C
Boiling Point 81°C
UN Number UN1145
Beilstein 1908842
Solubility Information Insoluble in water.
IUPAC Name 1,1,2,2,3,3,4,4,5,5,6,6-dodecadeuteriocyclohexane
Formula Weight 96.24
Percent Purity 99.5%
Chemical Name or Material Cyclohexane-d{12}, Isotopic
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.